Learn More
Enzo Life Sciences CD31 (human) monoclonal antibody (Gi18) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX805003AC100
Encompass_Preferred
Clone: Gi18. Isotype: IgG1. Immunogen: Human platelets. Species reactivity: Human. Specificity: Recognizes the Ig loops 1-2 of CD31 (PECAM-1). Applications: Flow Cytometry, IHC (FS), IHC (PS), WB. Formulation: Liquid. 1mg/ml in PBS, pH 7.2-7.4. Contains no preservatives.
Catalog No. 50-201-0028