Learn More
Biocare Medical, LLC CD31 PECAM-1 Antibody - Predilute, 6.0 ml
SDP
Supplier:  Biocare Medical, LLC PM131AA
Encompass_Preferred
It has been shown that CD31 PECAM-1 Antibody can detect vascular endothelium associated antigen and has been used as a marker for benign and malignant human vascular disorders, myeloid leukemia infiltrates and megakaryocytes in normal bone marrow. When compared to Factor VIII and CD34 antibodies, studies have shown CD31 to be a superior marker for angiogenesis; which reportedly predicts tumor recurrence. CD31 with CD34 and Factor VIII has been used to mark Kaposi sarcoma and angiosarcomas. Other studies indicate that CD31 and CD34 can be used as markers for myeloid progenitor cells that recognize different myeloid leukemia infiltrates.
Catalog No. 50-830-69