Learn More
Sino Biological Anti-CD31/PECAM-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50408T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CD31 / PECAM-1 (Catalog#50408-M08H; NP_033868.3; Met1-Lys590). CD31 / PECAM-1 specific IgG was purified by Mouse CD31 / PECAM-1 affinity chromatography.
Catalog No. 50-224-8202