Learn More
ACROBiosystems Recombinant Protein;Human CD31 / PECAM-1 Protein;HEK293;100UG;CD1,PECAM1,PECAM-1,CD31,CD-31,FLJ34100,FLJ58394,PECAM
Catalog No. 501657924
Encompass_Preferred
Recombinant Protein;Human CD31 / PECAM-1 Protein;100UG;Human CD31, His Tag (CD1-H5224) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Lys 601 (Accession # P16284-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 66.4 kDa. The protein migrates as 80-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7924 Supplier ACROBiosystems Supplier No. CD1H5224100UG
Recombinant Protein;Human CD31 / PECAM-1 Protein;100UG;Human CD31, His Tag (CD1-H5224) is expressed from human 293 cells (HEK293). It contains AA Gln 28 - Lys 601 (Accession # P16284-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 66.4 kDa. The protein migrates as 80-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.