Learn More
Sino Biological CD31/PECAM1 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 10148T60100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD31/PECAM1 (rh CD31/PECAM1; Catalog#10148-H08H; EAW94208.1; Met1-Lys601). CD31/PECAM1 specific IgG was purified by Human CD31/PECAM1 affinity chromatography.
Catalog No. 50-224-8088