Learn More
CAPRICO BIOTECHNOLOGIES INC CD32 Antibody, Clone 4027-045
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4027045
Encompass_Preferred
Caprico Biotechnologies' CD32 antibody is a mouse monoclonal antibody targeting human CD32 (FcγRIIb). Suitable for flow cytometry, immunohistochemistry, and Western blot, it comes in various sizes and formats like purified, FITC, and biotin conjugates. It's validated for these applications and aids in analyzing CD32's role in immune regulation.
Catalog No. 50-227-9114