Learn More
CAPRICO BIOTECHNOLOGIES INC Anti-CD326 FITC Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4096015
Encompass_Preferred
The CapricoBio Anti-CD326 FITC Antibody is a fluorescein isothiocyanate (FITC) conjugated monoclonal antibody that recognizes human CD326, also known as Epithelial Cell Adhesion Molecule (EpCAM). It is produced in mice and is suitable for flow cytometry, immunofluorescence microscopy, and immunohistochemistry. The antibody is supplied as 100 μg in 0.5 mL of PBS with 0.1% bovine serum albumin and 0.09% sodium azide. The immunogen is the human EpCAM extracellular domain. This antibody can be used for labeling and detecting CD326/EpCAM positive cells in various applications.
Catalog No. 50-227-9116