Learn More
CAPRICO BIOTECHNOLOGIES INC Anti-CD326 Antibody, iFluor 488
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4096115
Encompass_Preferred
The CapricoBio Anti-CD326 Antibody (IFluor 488) is a mouse monoclonal antibody that targets human CD326, also known as EpCAM. It is conjugated to IFluor 488 fluorescent dye and is suitable for flow cytometry, immunofluorescence, and immunohistochemistry applications. This antibody recognizes the extracellular domain of CD326 and has been tested for specificity in human cell lines. CD326 is a transmembrane glycoprotein expressed on epithelial cells and overexpressed in many carcinomas. Overall, this fluorescent antibody can be a useful research tool for analyzing CD326 expression and localization.
Catalog No. 50-227-9117