Learn More
ACROBiosystems Recombinant Protein;Human Fc gamma RIIA / CD32a (H167) Protein (SPR verified);HEK293;100UG;CD1,CD32a,FCGR2A,CD32,FCG2 ,FCGR2A1,IGFR2,Fcr,Fcgr,Fc receptor,Fc gamma receptor
Catalog No. 501017340
Encompass_Preferred
Recombinant Protein;Human Fc gamma RIIA / CD32a (H167) Protein (SPR verified);100UG;Human CD32a (H167) (SPR verified) (CD1-H5223) is expressed from human 293 cells (HEK293). It contains AA Ala 36 - Ile 218 (Accession # P12318-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.1 kDa. The protein migrates as 29-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD32a (H167) (Cat. No. CD1-H5223) on CM5 Chip via anti-His antibody, can bind  Rituximab with an affinity constant of 1.45 μM as determined in a SPR assay (Biacore T200) (QC tested).
Catalog No. 50-101-7340 Supplier ACROBiosystems Supplier No. CD1H5223100UG
Recombinant Protein;Human Fc gamma RIIA / CD32a (H167) Protein (SPR verified);100UG;Human CD32a (H167) (SPR verified) (CD1-H5223) is expressed from human 293 cells (HEK293). It contains AA Ala 36 - Ile 218 (Accession # P12318-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.1 kDa. The protein migrates as 29-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD32a (H167) (Cat. No. CD1-H5223) on CM5 Chip via anti-His antibody, can bind  Rituximab with an affinity constant of 1.45 μM as determined in a SPR assay (Biacore T200) (QC tested).