Learn More
Sino Biological Human CD32a / FCGR2A Protein (167 His, His Tag) 1mg
Supplier:  Sino Biological 10374H08C11
Encompass_Preferred
A DNA sequence encoding (Met1-Ile218) of human CD32a (P12318-1) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-211-5467