Learn More
BD Cell Analysis 3P CD33
Supplier:  BD Cell Analysis 3P 341640
Encompass
CD33, clone P67.6, is derived from the hybridization of mouse Sp2/0 myeloma cells with spleen cells from BALB/c mice immunized with FMY9S5 cells containing the CD33 gene. CD33 recognizes a human myelomonocytic antigen, Mr 67 kilodaltons (kd).
Catalog No. 50-620-524
Please call customer service for current pricing. 1-800-766-7000