Learn More
Aviva Systems Biology Corp CD33 Antibody - N-terminal region (ARP63067_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63067_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD33. It was validated on Western Blot. . Alternate names: FLJ00391, SIGLEC-3, SIGLEC3, p67ApplicationWB. Species reactivity: Cow, Horse, Human
Catalog No. 50-287-861