Learn More
CAPRICO BIOTECHNOLOGIES INC CD33 PE-Cyanine5, 40-4007-5
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4040075
Encompass_Preferred
The CD33 PE-Cyanine5 antibody by Caprico Biotechnologies is a mouse monoclonal that targets human CD33 for flow cytometry and more. It binds an extracellular CD33 epitope and is used to detect acute myeloid leukemia cells. Supplied in PBS with BSA and sodium azide, the recommended flow cytometry dilution is 5-25 μl/test. Optimal dilution should be determined by the user.
Catalog No. 50-227-9122