Learn More
CAPRICO BIOTECHNOLOGIES INC CD33 PerCP-Cyanine5.5, 40-40065
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4040065
Encompass_Preferred
The CD33 PerCP-Cyanine5.5 antibody from Caprico Biotechnologies is a mouse monoclonal antibody that recognizes human CD33. It is conjugated to PerCP-Cyanine5.5 and is suitable for flow cytometry applications. This antibody recognizes myeloid cells and acute myeloid leukemia blasts. It can be used for immunophenotyping and leukemia diagnosis. The clone is WM53. The antibody is supplied at 0.1 mg in PBS with 0.1% sodium azide and 2% BSA.
Catalog No. 50-227-9121