Learn More
Enzo Life Sciences CD33 Recombinant monoclonal antibody (hP67.6) (Human IgG4 kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6250200
Encompass_Preferred
Clone: hP67.6. Isotype: IgG4κ. Species reactivity: Human. Applications: Flow Cytometry, IF. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3149