Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 947MSM4P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: QBEnd/10 + HPCA1/763. Clonality: Monoclonal. Host: Mouse. Isotype: IgG. Positive Control: KG-1 cells, Tonsil, or Angiosarcoma. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Detergent solubilized vesicular suspension prepared from human term placenta (QBEnd/10); Recombinant full-length human HPCA1 protein (HPCA1/763). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 90-110kDa.Chromosome: 1q32.2. Gene Symbol: CD34. Gene ID: 947. Ensemble: ENSG00000174059. Uniprot: NP_001020280.
Catalog No. 50-188-5350