Learn More
Aviva Systems Biology Corp CD34 Antibody - C-terminal region (ARP60993_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60993_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD34. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CD34. Alternate names: -ApplicationIHC, WB. Species reactivity: Dog, Goat, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-226