Learn More
Aviva Systems Biology Corp CD34 Antibody - C-terminal region (ARP60994_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60994_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD34. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CD34. Alternate names: -ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-287-227