Learn More
Revvity Health Sciences Inc CD34+ HEMATOPOIETIC Stem cells
Catalog No. 502810724
Encompass
CD34+ cells are isolated from human umbilical cord blood using positive immunomagnetic separation techniques from a single or pooled cord blood unit(s) resulting in > 95% viability and > 90% purity. Certificate of Analysis is included. Cord Blood units are collected from donors following IRB-approved protocols and/or in accordance with local, state and national regulatory requirements. Vial, 1.8 mL, CD34+ cells frozen in 10% DMSO cryopreservation medium 1-million cells.
Catalog No. 50-281-0724 Supplier Revvity Health Sciences Inc Supplier No. CBCD341MS
CD34+ cells are isolated from human umbilical cord blood using positive immunomagnetic separation techniques from a single or pooled cord blood unit(s) resulting in > 95% viability and > 90% purity. Certificate of Analysis is included. Cord Blood units are collected from donors following IRB-approved protocols and/or in accordance with local, state and national regulatory requirements. Vial, 1.8 mL, CD34+ cells frozen in 10% DMSO cryopreservation medium 1-million cells.