Learn More
Genscript Corporation CD34 His Mouse
Catalog No. 5000057687
Encompass
CD34 is a transmembrane phosphoglycoprotein first identified on hematopoietic stem and progenitor cells Clinically it is associated with the selection and enrichment of hematopoietic stem cells for bone marrow transplants Due to these historical and clinical associations CD34 expression is almost ubiquitously related to hematopoietic cells and it is a common misconception that CD34-positive (CD34()) cells in nonhematopoietic samples represent hematopoietic contamination
Catalog No. 50-000-57687 Supplier Genscript Corporation Supplier No. Z04313500
CD34 is a transmembrane phosphoglycoprotein first identified on hematopoietic stem and progenitor cells Clinically it is associated with the selection and enrichment of hematopoietic stem cells for bone marrow transplants Due to these historical and clinical associations CD34 expression is almost ubiquitously related to hematopoietic cells and it is a common misconception that CD34-positive (CD34()) cells in nonhematopoietic samples represent hematopoietic contamination