Learn More
CAPRICO BIOTECHNOLOGIES INC CD34 iFluor700 Antibody
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4089195
Encompass_Preferred
The Caprico CD34 iFluor 700 Antibody is a mouse monoclonal antibody that specifically binds to human CD34. It is conjugated to iFluor 700 and is suitable for flow cytometry, immunohistochemistry, and immunofluorescence. This antibody recognizes CD34, a transmembrane phosphoglycoprotein expressed on hematopoietic stem and progenitor cells. It can be used to identify and isolate hematopoietic stem cells and is useful for research on hematopoiesis, leukemia, bone marrow transplantation, and stem cell isolation. The antibody is available in multiple sizes and concentrations for flexibility. Overall, this antibody enables detection and analysis of CD34+ cells via fluorescent labeling.
Catalog No. 50-227-9126