Learn More
OriGene CD34 MS MONOCLN ANTBY 100UG
SDP
Catalog No. NC4041944
Encompass_Preferred
CD34 Mouse Monoclonal Antibody 100UGClone name - EQ-8D11-C1Application-ELISA, FC, IF, WBReactivity-SheepAntibody Host-Mouse
Catalog No. NC4041944 Supplier OriGene Supplier No. AM32448PUN
CD34 Mouse Monoclonal Antibody 100UGClone name - EQ-8D11-C1Application-ELISA, FC, IF, WBReactivity-SheepAntibody Host-Mouse