Learn More
CAPRICO BIOTECHNOLOGIES INC CD34 Antibody, Clone 4H11, PE/CF594
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4089205
Encompass_Preferred
The CD34 Antibody (Clone 4H11) PE/CF594 is a mouse monoclonal antibody that binds to human CD34. CD34 is a transmembrane phosphoglycoprotein expressed on hematopoietic stem and progenitor cells. This antibody is conjugated to PE/CF594, a tandem fluorochrome with an emission maximum of 615nm, and is suitable for flow cytometry applications. It is available in multiple sizes ranging from 25 tests to 200 tests. This antibody is validated for various applications including Western Blot, Immunoprecipitation, and Flow Cytometry.
Catalog No. 50-227-9125