Learn More
Biogenex Laboratories CD34 : QBEnd/10 : 6mL
SDP
Supplier:  Biogenex Laboratories AM2365M
Encompass_Preferred
This is an antibody to the CD34 antigen in human endothelial and hematopoietic cells. It stains positive in a variety of vascular and lymphatic tumors. QBEnd/10 may now prove to be a more specific method of evaluating vascularization than Factor VIII antibody and is an important tool for tumor evaluation.
Catalog No. 50-197-8754