Learn More
Medchemexpress LLC CD34 Rat HEK293 F 10ug
SDP
Catalog No. 5000178602
Encompass_Preferred
CD34 Protein is a transmembrane phosphoglycoprotein that exists on the surface of human and various animal cells CD34 Protein is used as a cellular marker for hematopoietic stem cell sorting and as an islet endocrine cell marker CD34 Protein plays an important role in cell adhesion inflammatory cell chemotaxis cell proliferation and differentiation and enhancement of inflammatory response CD34 Protein Rat (HEK293 Fc) is the recombinant rat-derived CD34 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-78602 Supplier Medchemexpress LLC Supplier No. HYP7430410UG
CD34 Protein is a transmembrane phosphoglycoprotein that exists on the surface of human and various animal cells CD34 Protein is used as a cellular marker for hematopoietic stem cell sorting and as an islet endocrine cell marker CD34 Protein plays an important role in cell adhesion inflammatory cell chemotaxis cell proliferation and differentiation and enhancement of inflammatory response CD34 Protein Rat (HEK293 Fc) is the recombinant rat-derived CD34 protein expressed by HEK293 with C-hFc labeled tag