Learn More
Enzo Life Sciences CD34 Recombinant monoclonal antibody (HPCA1/1806R) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS3280100
Encompass_Preferred
Clone: HPCA1/1806R. Isotype: IgGκ. Immunogen: Recombinant full length human CD34. Species reactivity: Human, Rat. Applications: Flow Cytometry, IF, IHC (PS), WB. Purity: Protein A affinity purified. Formulation: Liquid. In 1X PBS containing 0.1 mg/ml BSA (US sourced) and 0.05% sodium azide.
Catalog No. 50-201-2859