Learn More
Enzo Life Sciences CD34 Recombinant monoclonal antibody (My10) (Mouse IgG1 kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6270200
Encompass_Preferred
Clone: My10. Isotype: IgG1κ. Immunogen: KG-1a Human myeloblastic leukemic cell line (immature human myeloid cell line) from a patient with nonlymphocytic leukemia. Species reactivity: Human. Applications: Flow Cytometry, IHC, IP. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3151