Learn More
Cayman Chemical CD34CTerm RabbIt Monclon 100uL
SDP
Catalog No. 502948476
Encompass_Preferred
A BET family protein inhibitor inhibits binding of BRD2 BRD3 and BRD4 to an acetylated histone H4 peptide in vitro decreases Myc levels in and inhibits proliferation of AML DLBCL and MM cells reduces tumor growth in AML DLBCL and MM mouse xenograft models reduces tumor growth when administered alone or in combination with docetaxel or MDV 3100 in a 22Rv1 CRPC mouse xenograft model
Catalog No. 50-294-8476 Supplier Cayman Chemical Supplier No. 32245100 UL
A BET family protein inhibitor inhibits binding of BRD2 BRD3 and BRD4 to an acetylated histone H4 peptide in vitro decreases Myc levels in and inhibits proliferation of AML DLBCL and MM cells reduces tumor growth in AML DLBCL and MM mouse xenograft models reduces tumor growth when administered alone or in combination with docetaxel or MDV 3100 in a 22Rv1 CRPC mouse xenograft model