Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 1378MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, IHC. Conjugate/Tag: Unconjugated. Clone: E11. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: Follicular dendritic cells (FDC) in tonsil. Cellular Staining: Cell surface. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Intact human monocytes. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 210-220kDa.Chromosome: 1q32.2. Gene Symbol: CR1. Gene ID: 1378. Ensemble: ENSG00000203710. Uniprot: NP_000642.
Catalog No. 50-188-4627