Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese Chloride, Tetrahydrate, USP, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18604187 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-187 50 kg Fiber Drum
18-604-183 125 g Fiber Drum
18-604-184 500 g Fiber Drum
18-604-185 2.5 kg Fiber Drum
18-604-186 12 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-187 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M110650KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

M1106, 13446-34-9, MnCl2·4H2O

Spectrum™ Chemical Manganese Chloride, Tetrahydrate, USP is indicated for use as a supplement to intravenous solutions given for total parenteral nutrition (TPN) to maintain Manganese serum levels. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Physical Form Fine Crystals or Crystalline Powder
CAS Max % 100%
CAS 7773-01-5
Quantity 50 kg
Loss on Drying 36 to 38.5%
Packaging Fiber Drum
Percent Purity 98 to 101%
Grade USP
Chemical Name or Material Manganese Chloride, Tetrahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.