Learn More
Enzo Life Sciences CD35 Recombinant monoclonal antibody (8C12) (Rat IgG2a kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS4360200
Encompass_Preferred
Clone: 8C12. Isotype: IgG2aκ. Immunogen: Purified murine CR1 protein. Species reactivity: Mouse. Specificity: Recognizes an epitope on CD35 (CR1, 190 kDa), but not on CD21 (CR2, 145-150 kDa), which are alternative splice variants of primary Cr2 gene transcripts. Applications: Flow Cytometry, IHC (FS), IP, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-2963