Learn More
Aviva Systems Biology Corp CD36 Antibody - N-terminal region (ARP48127_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48127_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CD36. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CD36. Alternate names: CHDS7, FAT, GP3B, GP4, GPIV, PASIV, SCARB3, BDPLT10ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59719