Learn More
ACROBiosystems ACROBiosystems Human CD36 / SR-B3 Protein, His Tag
Catalog No. 501957496
Encompass_Preferred
Recombinant Protein;500UG;Human CD36, His Tag (CD6-H5221) is expressed from human 293 cells (HEK293). It contains AA Gly 30 - Asn 439 (Accession # NP_001001547.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 48.5 kDa. The protein migrates as 66-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7496 Supplier ACROBiosystems Supplier No. CD6H5221500UG
Recombinant Protein;500UG;Human CD36, His Tag (CD6-H5221) is expressed from human 293 cells (HEK293). It contains AA Gly 30 - Asn 439 (Accession # NP_001001547.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 48.5 kDa. The protein migrates as 66-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.