Learn More
Enzo Life Sciences CD36 (human), (recombinant) (10µg)
SDP
Catalog No. 502013958
Encompass_Preferred
Alternative name: SCARB3, GP3B, GP4, Platelet glycoprotein 4. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from 0.22µm filtered solution in PBS, pH 7.4. Generally 5-8% mannitol or trehalose is added as a protectant before lyophilization. Source: Produced in HEK 293 cells. Long Term Storage: -20°C.
Catalog No. 50-201-3958 Supplier Enzo Life Sciences Supplier No. ENZPRT1700010
Alternative name: SCARB3, GP3B, GP4, Platelet glycoprotein 4. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from 0.22µm filtered solution in PBS, pH 7.4. Generally 5-8% mannitol or trehalose is added as a protectant before lyophilization. Source: Produced in HEK 293 cells. Long Term Storage: -20°C.