Learn More
Medchemexpress LLC CD36 Mouse HEK293 10ug
SDP
Catalog No. 5000181671
Encompass_Preferred
CD36 is a multifunctional glycoprotein that binds to proteins such as thrombospondin fibronectin collagen beta-amyloid and oxidized low-density lipoprotein as well as lipid molecules such as anionic phospholipids and long-chain phospholipids CD36 regulates angiogenesis inflammatory responses fatty acid metabolism taste dietary fat processing and acts as a TLR4 TLR6 heterodimer coreceptor to mediate inflammatory responses regulate immune responses and bacterial absorption CD36 Protein Mouse (HEK293 His) is the recombinant mouse-derived CD36 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-81671 Supplier Medchemexpress LLC Supplier No. HYP7289510UG
CD36 is a multifunctional glycoprotein that binds to proteins such as thrombospondin fibronectin collagen beta-amyloid and oxidized low-density lipoprotein as well as lipid molecules such as anionic phospholipids and long-chain phospholipids CD36 regulates angiogenesis inflammatory responses fatty acid metabolism taste dietary fat processing and acts as a TLR4 TLR6 heterodimer coreceptor to mediate inflammatory responses regulate immune responses and bacterial absorption CD36 Protein Mouse (HEK293 His) is the recombinant mouse-derived CD36 protein expressed by HEK293 with C-His labeled tag