Learn More
Medchemexpress LLC CD36 Mouse HEK293 10ug
SDP
Catalog No. 5000182444
Encompass_Preferred
CD36 is a multifunctional glycoprotein that binds to proteins such as thrombospondin fibronectin collagen beta-amyloid and oxidized low-density lipoprotein as well as lipid molecules such as anionic phospholipids and long-chain phospholipids CD36 regulates angiogenesis inflammatory responses fatty acid metabolism taste dietary fat processing and acts as a TLR4 TLR6 heterodimer coreceptor to mediate inflammatory responses regulate immune responses and bacterial absorption CD36 Protein Mouse (HEK293 Fc) is the recombinant mouse-derived CD36 protein expressed by HEK293 with C-hFc labeled tag
Catalog No. 50-001-82444 Supplier Medchemexpress LLC Supplier No. HYP7273310UG
CD36 is a multifunctional glycoprotein that binds to proteins such as thrombospondin fibronectin collagen beta-amyloid and oxidized low-density lipoprotein as well as lipid molecules such as anionic phospholipids and long-chain phospholipids CD36 regulates angiogenesis inflammatory responses fatty acid metabolism taste dietary fat processing and acts as a TLR4 TLR6 heterodimer coreceptor to mediate inflammatory responses regulate immune responses and bacterial absorption CD36 Protein Mouse (HEK293 Fc) is the recombinant mouse-derived CD36 protein expressed by HEK293 with C-hFc labeled tag