Learn More
CAPRICO BIOTECHNOLOGIES INC CD38 Antibody, APC Conjugated, 4008045
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4008045
Encompass_Preferred
The Caprico CD38 Antibody APC Conjugated is a monoclonal antibody that specifically binds to human CD38. It is conjugated to APC (allophycocyanin) and is designed for flow cytometry applications. This antibody recognizes both membrane-bound and soluble forms of CD38. It can be used for immunophenotyping of hematopoietic cells as CD38 is expressed on early hematopoietic cells, activated B and T cells, plasma cells, and myeloid cells. The antibody is supplied as 100 tests in PBS with 0.1% sodium azide and 0.2% BSA. Each vial contains 100μg of antibody.
Catalog No. 50-227-9132