Learn More
CAPRICO BIOTECHNOLOGIES INC CD38 Antibody APC-Cyanine7, 4008095
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4008095
Encompass_Preferred
The Caprico CD38 Antibody APC-Cyanine7 is a monoclonal antibody that binds to human CD38. It is conjugated to APC-Cyanine7 and is suitable for flow cytometry, immunohistochemistry, and immunofluorescence. This antibody recognizes an extracellular epitope and does not cross-react with mouse CD38. It can be used to identify CD38 expressing cells including plasma cells, activated T cells, NK cells, and myeloid cells. The APC-Cyanine7 fluorochrome has an excitation max of 743nm and emission max of 767nm. This antibody is available in sizes ranging from 25ug to 1mg.
Catalog No. 50-227-9142