Learn More
TRIBIOSCIENCE INC Cluster of Differentiation 38 (CD38 Cyclase) Activity Assay Kit (Fluorometric), 100 Tests
SDP
Catalog No. NC2886955
Encompass_Preferred
CD38 Assay Kit is a simple and fast fluorometric assay that measures the cyclase activity of CD38 in biological samples. CD38 catalyzes the conversion of nicotinamide guanine dinucleotide (NGD+) to cyclic GDP-ribose (cGDPR) which can be measured by fluorometric method at Ex/Em=300 nm/410 nm.
Catalog No. NC2886955 Supplier TRIBIOSCIENCE INC Supplier No. TBS2100100
CD38 Assay Kit is a simple and fast fluorometric assay that measures the cyclase activity of CD38 in biological samples. CD38 catalyzes the conversion of nicotinamide guanine dinucleotide (NGD+) to cyclic GDP-ribose (cGDPR) which can be measured by fluorometric method at Ex/Em=300 nm/410 nm.