Learn More
Enzo Life Sciences CD38 (human) monoclonal antibody (HB7) (Alexa Fluor 546) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS1580100
Encompass_Preferred
Clone: HB7. Isotype: IgG1. Immunogen: BJAB human B cell line. Species reactivity: Human. Applications: Flow Cytometry. Purity: Purified using QMA Hyper D ion exchange chromatography. Formulation: Liquid. In PBS containing 50% glycerol.
Catalog No. 50-201-2716