Learn More
Genscript Corporation CD38 His Cynomolgus
Supplier:  Genscript Corporation Z04119500
Encompass
CD38 (cluster of differentiation 38) also known as cyclic ADP ribose hydrolase is a transmembrane glycoprotein found on the surface of some immune cells including plasma cells activated or immature T and B cells monocytes and natural killer cells CD38 participates in cell adhesion signal transduction and calcium signaling
Catalog No. 50-000-58284