Learn More
TRIBIOSCIENCE INC CD38 hydrolase Activity Fluorometric Assay
SDP
Catalog No. NC3159876
Encompass
Tribioscience’s CD38 hydrolase Assay Kit is a simple and fast fluorometric assay that measures the hydrolase activity of CD38 in biological samples. CD38 catalyzes the conversion of substrate to a florescent product which can be measured by fluorometric method at Ex/Em=300 nm/410 nm.
Catalog No. NC3159876 Supplier TRIBIOSCIENCE INC Supplier No. TBS2047
Tribioscience’s CD38 hydrolase Assay Kit is a simple and fast fluorometric assay that measures the hydrolase activity of CD38 in biological samples. CD38 catalyzes the conversion of substrate to a florescent product which can be measured by fluorometric method at Ex/Em=300 nm/410 nm.