Learn More
Medchemexpress LLC HY-123999 10mg , CD38 inhibitor 1 CAS:1700637-55-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12399910MG
Encompass_Preferred
HY-123999 10mg CD38 inhibitor 1 CAS: 1700637-55-3 CD38 inhibitor 1 (compound 78c) is a potent CD38 inhibitor with IC50s of 7.3 nM and 1.9 nM for hCD38 and mouse CD38. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0624