Learn More
Medchemexpress LLC CD38 inhibitor 1 | 1700637-55-3 | MFCD32062812 | 99.3% | 413.53 | C22H27N3O3S | 250MG
SDP
Catalog No. 5000238057
Encompass_Preferred
CD38 inhibitor 1 (compound 78c) is a small-molecule inhibitor of CD38 with low-nanomolar potency against human and mouse CD38 (IC50 7.3 nM and 1.9 nM, respectively). Supplied as a solid research compound, it has the chemical formula C22H27N3O3S, a molecular weight of 413.53, and typical purity of about 99.3%. It is provided in multiple package sizes and includes storage guidance for powder and solution forms.
  • Low-nanomolar potency against human and mouse CD38 (IC50 7.3 nM; 1.9 nM)
  • High chemical purity suitable for research applications
  • Available in multiple package sizes including 250 mg
  • Solid, light yellow to brown appearance for easy handling
  • Stable as powder at -20°C for long-term storage and at 4°C for shorter-term storage
  • Suitable for biochemical and cellular CD38 activity assays
Catalog No. 50-002-38057 Supplier Medchemexpress LLC Supplier No. HY123999250MG
CD38 inhibitor 1 (compound 78c) is a small-molecule inhibitor of CD38 with low-nanomolar potency against human and mouse CD38 (IC50 7.3 nM and 1.9 nM, respectively). Supplied as a solid research compound, it has the chemical formula C22H27N3O3S, a molecular weight of 413.53, and typical purity of about 99.3%. It is provided in multiple package sizes and includes storage guidance for powder and solution forms.
  • Low-nanomolar potency against human and mouse CD38 (IC50 7.3 nM; 1.9 nM)
  • High chemical purity suitable for research applications
  • Available in multiple package sizes including 250 mg
  • Solid, light yellow to brown appearance for easy handling
  • Stable as powder at -20°C for long-term storage and at 4°C for shorter-term storage
  • Suitable for biochemical and cellular CD38 activity assays