Learn More
Medchemexpress LLC CD38 inhibitor 1 (compound 78c) | 1700637-55-3 | 99.64% | 413.53 | 5 MG
SDP
Catalog No. 5000364598
Encompass_Preferred
CD38 inhibitor 1 (compound 78c) is a potent inhibitor of CD38, with IC50 values of 7.3 nM for human CD38 and 1.9 nM for mouse CD38. It also has a Ki of 0.3 nM for wild-type human CD38. In vivo studies showed that a 30 mg/kg oral administration of CD38 inhibitor 1 significantly increased NAD levels in the liver and muscle of diet-induced obese C57Bl6 mice at both 2 and 6-hour time points.
  • Potent CD38 inhibitor with low IC50 values for both human and mouse CD38.
  • Elevates NAD levels in liver and muscle in vivo.
Catalog No. 50-003-64598 Supplier Medchemexpress LLC Supplier No. 5MGHY123999
CD38 inhibitor 1 (compound 78c) is a potent inhibitor of CD38, with IC50 values of 7.3 nM for human CD38 and 1.9 nM for mouse CD38. It also has a Ki of 0.3 nM for wild-type human CD38. In vivo studies showed that a 30 mg/kg oral administration of CD38 inhibitor 1 significantly increased NAD levels in the liver and muscle of diet-induced obese C57Bl6 mice at both 2 and 6-hour time points.
  • Potent CD38 inhibitor with low IC50 values for both human and mouse CD38.
  • Elevates NAD levels in liver and muscle in vivo.