Learn More
Selleck Chemical LLC CD38 inhibitor 15mg
SDP
Supplier:  Selleck Chemical LLC S89605MG
Encompass_Preferred
CD38 inhibitor 1 (compound 78c, CD38-IN-78c, MDK-7553) is a potent inhibitor of CD38 with IC50 of 7.3 nM and 1.9 nM for human CD38 and mouse CD38, respetively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3458