Learn More
Medchemexpress LLC CD38 inhibitor 2 | 2597933-78-1 | 98.8% | 384.43 g/mol | C19H24N6O3 | 5 MG
SDP
Catalog No. 5000221420
Encompass_Preferred
CD38 inhibitor 2 is a small-molecule inhibitor of the CD38 enzyme with submicromolar potency (IC50 = 0.01-0.1 μM). Provided as a solid research compound and as a 10 mM solution in DMSO, it is intended for in vitro biochemical and cellular studies of CD38-related biology. The compound has formula C19H24N6O3, molecular weight 384.43 g/mol, and is supplied at high purity.
  • Potent CD38 inhibition (IC50 0.01-0.1 μM).
  • High purity (~98.8%) suitable for biochemical and cellular assays.
  • Available as a solid and as a 10 mM solution in DMSO for convenience.
  • Supplied in small research quantities (for example, 5 mg) for screening and profiling.
  • Molecular formula C19H24N6O3 and molecular weight 384.43 g/mol for experimental planning.
Catalog No. 50-002-21420 Supplier Medchemexpress LLC Supplier No. HY1449885MG
CD38 inhibitor 2 is a small-molecule inhibitor of the CD38 enzyme with submicromolar potency (IC50 = 0.01-0.1 μM). Provided as a solid research compound and as a 10 mM solution in DMSO, it is intended for in vitro biochemical and cellular studies of CD38-related biology. The compound has formula C19H24N6O3, molecular weight 384.43 g/mol, and is supplied at high purity.
  • Potent CD38 inhibition (IC50 0.01-0.1 μM).
  • High purity (~98.8%) suitable for biochemical and cellular assays.
  • Available as a solid and as a 10 mM solution in DMSO for convenience.
  • Supplied in small research quantities (for example, 5 mg) for screening and profiling.
  • Molecular formula C19H24N6O3 and molecular weight 384.43 g/mol for experimental planning.