Learn More
BPS Bioscience Inc CD38 INHIBITOR SCREENING ASSAY
SDP
Catalog No. NC1563847
Encompass
CD38 Inhibitor Screening Assay Kit (Hydrolase Activity)The CD38 Inhibitor Screening Assay Kit (Hydrolase Activity) is designed to measure the glycohydrolase activity of CD38 for screening and profiling applications. The CD38 assay kit comes in a convenient 96-well format, with purified recombinant CD38 enzyme, its substrate N6-etheno-NAD (-NAD), and CD38 assay buffer for 100 enzyme reactions. In addition, the kit includes the CD38 inhibitor apigenin for use as a control inhibitor.
Catalog No. NC1563847 Supplier BPS Bioscience Inc Supplier No. 79287
CD38 Inhibitor Screening Assay Kit (Hydrolase Activity)The CD38 Inhibitor Screening Assay Kit (Hydrolase Activity) is designed to measure the glycohydrolase activity of CD38 for screening and profiling applications. The CD38 assay kit comes in a convenient 96-well format, with purified recombinant CD38 enzyme, its substrate N6-etheno-NAD (-NAD), and CD38 assay buffer for 100 enzyme reactions. In addition, the kit includes the CD38 inhibitor apigenin for use as a control inhibitor.