Learn More
CAPRICO BIOTECHNOLOGIES INC CD38 PE-Cyanine7, 4008085
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4008085
Encompass_Preferred
The Caprico CD38 PE-Cyanine7 antibody is a mouse monoclonal antibody that recognizes human CD38. It is conjugated to PE-Cyanine7 and is suitable for flow cytometry applications. This antibody detects CD38 expression on various hematopoietic cells including plasma cells, activated T and B lymphocytes, NK cells, monocytes, macrophages, granulocytes, and platelets. It can be used for immunophenotyping and to identify leukemic cells. The antibody is supplied as 100 tests in PBS with 0.1% sodium azide and 0.2% BSA.
Catalog No. 50-227-9143