Learn More
CAPRICO BIOTECHNOLOGIES INC CD38 PerCP Antibody, 4008035
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4008035
Encompass_Preferred
The Caprico CD38 PerCP Antibody is a mouse monoclonal antibody that specifically binds to human CD38. It is conjugated to PerCP and is suitable for flow cytometry applications. This antibody recognizes an epitope on the ecto-enzyme domain of CD38 and does not block the enzymatic activity. CD38 is expressed on hematopoietic cells including T, B, NK cells, monocytes, macrophages and granulocytes. This antibody can be used to identify and isolate CD38 expressing cells by flow cytometry.
Catalog No. 50-227-9139